Assay Details
Gene Symbol: LPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101377.5 | NP_172961.1 | 3 - 4 | 586 | 129 | |
GenBank mRNA | AB053950.1 | - | 2 - 3 | 304 | 129 | |
AB061407.1 | - | 3 - 4 | 507 | 129 | ||
AY087673.1 | - | 3 - 4 | 499 | 129 | ||
BT003912.1 | - | 3 - 4 | 499 | 129 | ||
BT004969.1 | - | 2 - 3 | 304 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
