Assay Details
Gene Symbol: LPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101377.5 | NP_172961.1 | 5 - 6 | 796 | 99 | |
GenBank mRNA | AB053950.1 | - | 4 - 5 | 514 | 99 | |
AB061407.1 | - | 5 - 6 | 717 | 99 | ||
AY087673.1 | - | 5 - 6 | 709 | 99 | ||
BT003912.1 | - | 5 - 6 | 709 | 99 | ||
BT004969.1 | - | 4 - 5 | 514 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
