Assay Details
Gene Symbol: LPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101377.5 | NP_172961.1 | 7 - 8 | 1040 | 83 | |
GenBank mRNA | AB053950.1 | - | 6 - 7 | 758 | 83 | |
AB061407.1 | - | 7 - 8 | 961 | 83 | ||
AY087673.1 | - | 7 - 8 | 953 | 83 | ||
BT003912.1 | - | 7 - 8 | 953 | 83 | ||
BT004969.1 | - | 6 - 7 | 758 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
