Assay Details
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332212.1 | NP_001323115.1 | 1 - 2 | 626 | 60 | |
NM_101486.3 | NP_173070.1 | 2 - 3 | 380 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
