Assay Details
Gene Symbol: MYB58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101514.3 | NP_173098.1 | 1 - 2 | 382 | 119 | |
GenBank mRNA | AB493459.1 | - | 1 - 2 | 138 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
