Assay Details
Gene Symbol: ABA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126103.2 | NP_201504.2 | 14 - 15 | 1970 | 95 | |
NM_180954.3 | NP_851285.1 | 14 - 15 | 2132 | 95 | ||
GenBank mRNA | AY093145.1 | - | 14 - 15 | 1900 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
