Assay Details
Gene Symbol: ABA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126103.2 | NP_201504.2 | 4 - 5 | 994 | 142 | |
NM_180954.3 | NP_851285.1 | 4 - 5 | 1160 | 142 | ||
GenBank mRNA | AY093145.1 | - | 4 - 5 | 928 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
