Assay Details
Gene Symbol: ABA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126103.2 | NP_201504.2 | 5 - 6 | 1098 | 89 | |
NM_180954.3 | NP_851285.1 | 5 - 6 | 1264 | 89 | ||
GenBank mRNA | AY093145.1 | - | 5 - 6 | 1032 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
