Assay Details
Gene Symbol: ABA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126103.2 | NP_201504.2 | 6 - 7 | 1323 | 83 | |
NM_180954.3 | NP_851285.1 | 6 - 7 | 1489 | 83 | ||
GenBank mRNA | AY093145.1 | - | 6 - 7 | 1257 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
