Assay Details
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126259.4 | NP_178307.2 | 12 - 13 | 1299 | 74 | |
GenBank mRNA | AY062746.1 | - | 12 - 13 | 1291 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
