Assay Details
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126259.4 | NP_178307.2 | 14 - 15 | 1448 | 88 | |
GenBank mRNA | AY062746.1 | - | 14 - 15 | 1440 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
