Assay Details
Gene Symbol: SOS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126259.4 | NP_178307.2 | 16 - 17 | 1977 | 104 | |
GenBank mRNA | AY062746.1 | - | 16 - 17 | 1969 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
