Assay Details
Gene Symbol: LPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084628.2 | NP_001078097.1 | 2 - 3 | 739 | 81 | |
NM_001337419.1 | NP_001327801.1 | 2 - 3 | 1220 | 81 | ||
NM_001337421.1 | NP_001319453.1 | 2 - 3 | 306 | 81 | ||
NM_001337425.1 | NP_001327799.1 | 1 - 2 | 1340 | 81 | ||
NM_111128.2 | NP_566177.1 | 2 - 3 | 495 | 81 | ||
GenBank mRNA | AY084915.1 | - | 2 - 3 | 328 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001084628.2, NM_001337419.1, NM_001337421.1, NM_001337425.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
