Assay Details
Gene Symbol: LPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084626.2 | NP_001078095.1 | 4 - 5 | 815 | 87 | |
NM_001084627.1 | NP_001078096.1 | 4 - 5 | 528 | 87 | ||
NM_001084628.2 | NP_001078097.1 | 5 - 6 | 1152 | 87 | ||
NM_001084629.2 | NP_001078098.1 | 4 - 5 | 640 | 87 | ||
NM_001337419.1 | NP_001327801.1 | 5 - 6 | 1633 | 87 | ||
NM_001337420.1 | NP_001327802.1 | 3 - 4 | 2142 | 87 | ||
NM_001337421.1 | NP_001319453.1 | 5 - 6 | 719 | 87 | ||
NM_001337422.1 | NP_001327800.1 | 4 - 5 | 1078 | 87 | ||
NM_001337423.1 | NP_001327797.1 | 5 - 6 | 958 | 87 | ||
NM_001337424.1 | NP_001327798.1 | 4 - 5 | 2034 | 87 | ||
NM_001337425.1 | NP_001327799.1 | 4 - 5 | 1753 | 87 | ||
NM_111128.2 | NP_566177.1 | 5 - 6 | 908 | 87 | ||
GenBank mRNA | AY084915.1 | - | 5 - 6 | 741 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
