Assay Details
Gene Symbol: AT3G03590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111230.4 | NP_566210.1 | 1 - 2 | 642 | 86 | |
GenBank mRNA | AK117771.1 | - | 1 - 2 | 642 | 86 | |
BT003701.1 | - | 1 - 2 | 303 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
