Assay Details
Gene Symbol: CS26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111234.5 | NP_187013.1 | 4 - 5 | 804 | 154 | |
GenBank mRNA | AB003041.1 | - | 4 - 5 | 783 | 154 | |
AY099573.1 | - | 4 - 5 | 789 | 154 | ||
BT002155.1 | - | 4 - 5 | 743 | 154 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
