Assay Details
Gene Symbol: EDA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111236.3 | NP_187015.1 | 1 - 2 | 942 | 83 | |
GenBank mRNA | AK228299.1 | - | 1 - 2 | 942 | 83 | |
BT006471.1 | - | 1 - 2 | 698 | 83 | ||
KJ138843.1 | - | 1 - 2 | 698 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
