Assay Details
Gene Symbol: EDA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111236.3 | NP_187015.1 | 2 - 3 | 1234 | 115 | |
GenBank mRNA | AK228299.1 | - | 2 - 3 | 1234 | 115 | |
BT006471.1 | - | 2 - 3 | 990 | 115 | ||
KJ138843.1 | - | 2 - 3 | 990 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
