Assay Details
Gene Symbol: LSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036601.1 | NP_001031678.1 | 4 - 5 | 396 | 61 | |
NM_001036602.2 | NP_001031679.1 | 4 - 5 | 413 | 61 | ||
NM_001036603.2 | NP_001031680.2 | 4 - 5 | 496 | 61 | ||
NM_001084944.2 | NP_001078413.1 | 3 - 4 | 588 | 61 | ||
NM_001341436.1 | NP_001328264.1 | 4 - 5 | 507 | 61 | ||
NM_118157.5 | NP_567599.3 | 4 - 5 | 464 | 61 | ||
NM_179217.4 | NP_849548.1 | 4 - 5 | 507 | 61 | ||
NM_179218.4 | NP_849549.1 | 4 - 5 | 540 | 61 | ||
GenBank mRNA | AY080824.1 | - | 4 - 5 | 415 | 61 | |
AY087794.1 | - | 4 - 5 | 493 | 61 | ||
AY117316.1 | - | 2 - 3 | 228 | 61 | ||
U87833.1 | - | 4 - 5 | 454 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
