Assay Details
Gene Symbol: LSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036601.1 | NP_001031678.1 | 5 - 6 | 537 | 79 | |
NM_001036602.2 | NP_001031679.1 | 5 - 6 | 554 | 79 | ||
NM_001036603.2 | NP_001031680.2 | 5 - 6 | 637 | 79 | ||
NM_001084944.2 | NP_001078413.1 | 4 - 5 | 729 | 79 | ||
NM_001160785.1 | NP_001154257.1 | 7 - 8 | 712 | 79 | ||
NM_118157.5 | NP_567599.3 | 5 - 6 | 605 | 79 | ||
NM_179217.4 | NP_849548.1 | 5 - 6 | 648 | 79 | ||
NM_179218.4 | NP_849549.1 | 5 - 6 | 681 | 79 | ||
GenBank mRNA | AY080824.1 | - | 5 - 6 | 556 | 79 | |
AY087794.1 | - | 5 - 6 | 634 | 79 | ||
AY117316.1 | - | 3 - 4 | 369 | 79 | ||
U87833.1 | - | 5 - 6 | 595 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
