Assay Details
Gene Symbol: LSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036601.1 | NP_001031678.1 | 7 - 8 | 685 | 127 | |
NM_001036602.2 | NP_001031679.1 | 7 - 8 | 702 | 127 | ||
NM_001036603.2 | NP_001031680.2 | 7 - 8 | 785 | 127 | ||
NM_001084944.2 | NP_001078413.1 | 6 - 7 | 877 | 127 | ||
NM_001160785.1 | NP_001154257.1 | 9 - 10 | 860 | 127 | ||
NM_001341436.1 | NP_001328264.1 | 6 - 7 | 896 | 127 | ||
NM_118157.5 | NP_567599.3 | 7 - 8 | 753 | 127 | ||
NM_179217.4 | NP_849548.1 | 7 - 8 | 796 | 127 | ||
NM_179218.4 | NP_849549.1 | 7 - 8 | 829 | 127 | ||
GenBank mRNA | AY080824.1 | - | 7 - 8 | 704 | 127 | |
AY087794.1 | - | 7 - 8 | 782 | 127 | ||
U87833.1 | - | 7 - 8 | 743 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
