Assay Details
Gene Symbol: PPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203886.1 | NP_001190815.1 | 2 - 3 | 506 | 110 | |
NM_001341624.1 | NP_001320050.1 | 1 - 2 | 823 | 110 | ||
NM_001341625.1 | NP_001328939.1 | 2 - 3 | 564 | 110 | ||
NM_118497.4 | NP_567688.1 | 1 - 2 | 845 | 110 | ||
NM_179100.4 | NP_849431.1 | 2 - 3 | 591 | 110 | ||
GenBank mRNA | AB052553.2 | - | 1 - 2 | 643 | 110 | |
AY059864.1 | - | 2 - 3 | 490 | 110 | ||
BT008496.1 | - | 1 - 2 | 445 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
