Assay Details
Gene Symbol: AT1G31790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102913.3 | NP_174459.1 | 1 | 1151 | 149 | |
GenBank mRNA | BT026440.1 | - | 1 | 1106 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
