Assay Details
Gene Symbol: NSI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084172.1 | NP_001077641.1 | 3 - 4 | 203 | 81 | |
NM_102941.3 | NP_564387.1 | 3 - 4 | 321 | 81 | ||
NM_202221.2 | NP_973950.1 | 3 - 4 | 225 | 81 | ||
GenBank mRNA | AY086183.1 | - | 3 - 4 | 189 | 81 | |
AY281105.1 | - | 3 - 4 | 225 | 81 | ||
BT024481.1 | - | 3 - 4 | 173 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
