Assay Details
Gene Symbol: NSI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102941.3 | NP_564387.1 | 5 - 6 | 633 | 122 | |
NM_202221.2 | NP_973950.1 | 5 - 6 | 537 | 122 | ||
GenBank mRNA | AY086183.1 | - | 5 - 6 | 501 | 122 | |
AY281105.1 | - | 5 - 6 | 537 | 122 | ||
BT024481.1 | - | 5 - 6 | 485 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
