Assay Details
Gene Symbol: KAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342508.1 | NP_001329354.1 | 4 - 5 | 1702 | 66 | |
NM_120021.2 | NP_195572.2 | 4 - 5 | 1254 | 66 | ||
NM_179236.3 | NP_849567.2 | 4 - 5 | 1702 | 66 | ||
GenBank mRNA | AY265959.1 | - | 4 - 5 | 1294 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
