Assay Details
Gene Symbol: UGT73B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127109.3 | NP_179151.2 | 1 - 2 | 1102 | 95 | |
GenBank mRNA | AK227684.1 | - | 1 - 2 | 1102 | 95 | |
BT008319.1 | - | 1 - 2 | 987 | 95 | ||
KJ138913.1 | - | 1 - 2 | 987 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
