Assay Details
Gene Symbol: AT2G47280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180140.2 | NP_850471.2 | 3 - 4 | 607 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
