Assay Details
Gene Symbol: SNRK2.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120165.2 | NP_195711.1 | 4 - 5 | 480 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
