Assay Details
Gene Symbol: MYB3R-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203277.2 | NP_001190206.1 | 7 - 8 | 760 | 116 | |
NM_001342653.1 | NP_001331277.1 | 5 - 6 | 466 | 116 | ||
NM_120310.2 | NP_568099.1 | 7 - 8 | 721 | 116 | ||
GenBank mRNA | AY519649.1 | - | 7 - 8 | 634 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
