Assay Details
Gene Symbol: GSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342670.1 | NP_001318461.1 | 6 - 7 | 563 | 96 | |
NM_120356.4 | NP_195898.2 | 5 - 6 | 556 | 96 | ||
GenBank mRNA | BT010718.1 | - | 5 - 6 | 531 | 96 | |
BT012424.1 | - | 5 - 6 | 450 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
