Assay Details
Gene Symbol: GSTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125685.1 | NP_001119157.1 | 7 - 8 | 704 | 64 | |
NM_001342670.1 | NP_001318461.1 | 8 - 9 | 742 | 64 | ||
NM_120356.4 | NP_195898.2 | 7 - 8 | 735 | 64 | ||
GenBank mRNA | BT010718.1 | - | 7 - 8 | 710 | 64 | |
BT012424.1 | - | 7 - 8 | 629 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
