Assay Details
Gene Symbol: AT5G03610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120442.4 | NP_195981.1 | 1 - 2 | 328 | 117 | |
GenBank mRNA | AF360329.1 | - | 1 - 2 | 198 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
