Assay Details
Gene Symbol: AT5G03610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120442.4 | NP_195981.1 | 2 - 3 | 535 | 105 | |
GenBank mRNA | AF360329.1 | - | 2 - 3 | 405 | 105 | |
AY085113.1 | - | 2 - 3 | 304 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
