Assay Details
Gene Symbol: AT5G03610 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120442.4 | NP_195981.1 | 4 - 5 | 1123 | 134 | |
GenBank mRNA | AF360329.1 | - | 4 - 5 | 993 | 134 | |
AY085113.1 | - | 4 - 5 | 892 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
