Assay Details
Gene Symbol: AT2G18520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127408.2 | NP_565439.1 | 1 | 1476 | 77 | |
GenBank mRNA | AY088617.1 | - | 1 | 1428 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
