Assay Details
Gene Symbol: AT2G18940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127451.2 | NP_179484.1 | 1 | 2541 | 92 | |
GenBank mRNA | AY056798.1 | - | 1 | 2458 | 92 | |
BT010749.1 | - | 1 | 2404 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
