Assay Details
Gene Symbol: AT2G18980 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127455.4 | NP_179488.1 | 1 - 2 | 354 | 61 | |
GenBank mRNA | AK227636.1 | - | 1 - 2 | 226 | 61 | |
BT003082.1 | - | 1 - 2 | 208 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
