Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127520.4 | NP_179552.1 | 1 - 2 | 372 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
