Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084449.2 | NP_001077918.1 | 2 - 3 | 364 | 86 | |
NM_001335640.1 | NP_001323956.1 | 1 - 2 | 190 | 86 | ||
NM_127520.4 | NP_179552.1 | 2 - 3 | 466 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001335640.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
