Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084449.2 | NP_001077918.1 | 3 - 4 | 469 | 104 | |
NM_001335640.1 | NP_001323956.1 | 2 - 3 | 295 | 104 | ||
NM_127520.4 | NP_179552.1 | 3 - 4 | 571 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
