Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084449.2 | NP_001077918.1 | 8 - 9 | 839 | 81 | |
NM_001335640.1 | NP_001323956.1 | 7 - 8 | 665 | 81 | ||
NM_127520.4 | NP_179552.1 | 8 - 9 | 941 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
