Assay Details
Gene Symbol: NDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084449.2 | NP_001077918.1 | 9 - 10 | 1035 | 72 | |
NM_001335640.1 | NP_001323956.1 | 8 - 9 | 861 | 72 | ||
NM_127520.4 | NP_179552.1 | 9 - 10 | 1137 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
