Assay Details
Gene Symbol: PRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127534.3 | NP_179566.1 | 2 - 3 | 392 | 92 | |
GenBank mRNA | AY072427.1 | - | 2 - 3 | 349 | 92 | |
BT000264.1 | - | 2 - 3 | 265 | 92 | ||
U43325.1 | - | 2 - 3 | 365 | 92 | ||
U43590.1 | - | 2 - 3 | 341 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
