Assay Details
Gene Symbol: RPN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127618.4 | NP_565477.1 | 19 - 20 | 2477 | 89 | |
GenBank mRNA | AF360216.1 | - | 19 - 20 | 2429 | 89 | |
AY040061.1 | - | 19 - 20 | 2228 | 89 | ||
AY230828.1 | - | 19 - 20 | 2312 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
