Assay Details
Gene Symbol: AT2G20920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127661.4 | NP_179688.1 | 1 - 2 | 613 | 90 | |
GenBank mRNA | AY069895.1 | - | 1 - 2 | 373 | 90 | |
AY124817.1 | - | 1 - 2 | 335 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
