Assay Details
Gene Symbol: AT2G20920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127661.4 | NP_179688.1 | 2 - 3 | 966 | 76 | |
GenBank mRNA | AY069895.1 | - | 2 - 3 | 726 | 76 | |
AY124817.1 | - | 2 - 3 | 688 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
