Assay Details
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 19 - 20 | 2030 | 62 | |
GenBank mRNA | AY136413.1 | - | 19 - 20 | 1946 | 62 | |
BT012191.1 | - | 19 - 20 | 1889 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
