Assay Details
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 1 - 2 | 381 | 72 | |
GenBank mRNA | AY136413.1 | - | 1 - 2 | 297 | 72 | |
BT012191.1 | - | 1 - 2 | 240 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
