Assay Details
Gene Symbol: MYB33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085068.2 | NP_001078537.1 | 3 - 4 | 1692 | 92 | |
NM_001342848.1 | NP_001330997.1 | 2 - 3 | 1968 | 92 | ||
NM_001342849.1 | NP_001330996.1 | 3 - 4 | 1869 | 92 | ||
NM_180448.2 | NP_850779.1 | 4 - 5 | 1611 | 92 | ||
GenBank mRNA | AF411969.1 | - | 4 - 5 | 1437 | 92 | |
AK118937.1 | - | 4 - 5 | 1532 | 92 | ||
AY519616.1 | - | 2 - 3 | 1309 | 92 | ||
BT006049.1 | - | 2 - 3 | 1309 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
